siRNA Details
| Gene Symbol: Zscan22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001447.3 | NP_001001447.1 | 3 | 977 | ||
| NM_001290436.1 | NP_001277365.1 | 3 | 1014 | |||
| NM_001290438.1 | NP_001277367.1 | 3 | 911 | |||
| NM_001290440.1 | NP_001277369.1 | 2 | 426 | |||
| NM_001290441.1 | NP_001277370.1 | 2 | 492 | |||
| NM_001290442.1 | NP_001277371.1 | 2 | 529 | |||
| GenBank mRNA | AK052559.1 | - | 3 | 972 | ||
| AK078591.1 | - | 3 | 881 | |||
| AK085207.1 | - | 3 | 972 | |||
| AK163694.1 | - | 2 | 503 | |||
| BC026676.1 | - | 2 | 458 | 5053169 | ||

Zscan22
Genomic Map