siRNA Details
| Gene Symbol: Ccdc61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033314.3 | NP_001028486.1 | 1 | 236 | ||
| GenBank mRNA | AK082980.1 | - | 1 | 231 | ||
| AK146297.1 | - | 1 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc61| Gene Symbol: Ccdc61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033314.3 | NP_001028486.1 | 1 | 236 | ||
| GenBank mRNA | AK082980.1 | - | 1 | 231 | ||
| AK146297.1 | - | 1 | 237 | |||