siRNA Details
| Gene Symbol: Ppm1n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177691.3 | NP_808359.1 | 3, 4 | 1175 | ||
| GenBank mRNA | AK044232.1 | - | 3, 4 | 1175 | ||
| AK044334.1 | - | 3, 4 | 1176 | |||
| BC096372.1 | - | 3, 4 | 1164 | 4507828 | ||

Ppm1n
Genomic Map