siRNA Details
| Gene Symbol: Vmn1r178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206868.1 | NP_996751.1 | 1 | 808 | ||
| GenBank mRNA | BC154405.1 | - | 811 | 40050577 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r178| Gene Symbol: Vmn1r178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206868.1 | NP_996751.1 | 1 | 808 | ||
| GenBank mRNA | BC154405.1 | - | 811 | 40050577 | ||