siRNA Details
| Gene Symbol: Zfp114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029933.2 | NP_001025104.2 | 4 | 1109 | ||
| GenBank mRNA | AK139475.1 | - | 3 | 2028 | ||
| BC096771.1 | - | 2 | 1335 | |||
| BC138311.1 | - | 2 | 1224 | 8861331 | ||
| BC138312.1 | - | 2 | 1224 | 8861332 | ||

Zfp114
Genomic Map