siRNA Details
| Gene Symbol: Zfp428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290461.1 | NP_001277390.1 | 3 | 1170 | ||
| NM_146183.2 | NP_666295.2 | 3 | 1108 | |||
| NM_175142.1 | NP_780351.1 | 3 | 1147 | |||
| GenBank mRNA | AK010415.2 | - | 3 | 987 | ||
| AK080517.1 | - | 3 | 1101 | |||
| AK139174.1 | - | 3 | 839 | |||
| BC024802.1 | - | 3 | 1009 | 5360536 | ||
| BC096675.1 | - | 3 | 1021 | 5146693 | ||

Zfp428
Genomic Map