siRNA Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 924 | ||
| NM_146184.4 | NP_666296.1 | 1 | 1316 | |||
| GenBank mRNA | AK042388.1 | - | 1 | 1299 | ||
| AK154220.1 | - | 911 | ||||
| AK154901.1 | - | 2 | 880 | |||
| BC025206.1 | - | 2 | 910 | 5037295 | ||

B3gnt8
Genomic Map