siRNA Details
| Gene Symbol: Hnrnpul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144922.2 | NP_659171.1 | 2 | 470 | ||
| NM_178089.2 | NP_835190.1 | 2 | 451 | |||
| GenBank mRNA | AK042090.1 | - | 2 | 451 | ||
| AK087545.1 | - | 2 | 440 | |||
| AK143666.1 | - | 2 | 513 | |||
| AK155604.1 | - | 2 | 481 | |||
| AK202662.1 | - | 1 | 103 | |||
| BC021506.1 | - | 2 | 494 | 5347099 | ||
| BC078642.1 | - | 2 | 394 | 6329630 | ||

Hnrnpul1
Genomic Map