siRNA Details
| Gene Symbol: Blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290525.1 | NP_001277454.1 | 4 | 458 | ||
| NM_144923.3 | NP_659172.1 | 4 | 544 | |||
| GenBank mRNA | AK153153.1 | - | 4 | 465 | ||
| BC006617.1 | - | 4 | 484 | 3967600 | ||
| BC027279.1 | - | 4 | 544 | 3492861 | ||

Blvrb
Genomic Map