siRNA Details
Gene Symbol: Zfp850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254951.1 | NP_001241880.1 | 6 | 731 | ||
GenBank mRNA | CF735501.1 | - | 3 | 410 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254951.1 | NP_001241880.1 | 6 | 731 | ||
GenBank mRNA | CF735501.1 | - | 3 | 410 |