siRNA Details
| Gene Symbol: Samd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175021.3 | NP_778186.2 | 7 | 1422 | ||
| GenBank mRNA | BC042901.1 | - | 7 | 1475 | 5132736 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd4b| Gene Symbol: Samd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175021.3 | NP_778186.2 | 7 | 1422 | ||
| GenBank mRNA | BC042901.1 | - | 7 | 1475 | 5132736 | |