siRNA Details
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175021.3 | NP_778186.2 | 3 | 853 | ||
GenBank mRNA | BC042901.1 | - | 3 | 906 | 5132736 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175021.3 | NP_778186.2 | 3 | 853 | ||
GenBank mRNA | BC042901.1 | - | 3 | 906 | 5132736 |