siRNA Details
| Gene Symbol: Zfp940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173738.2 | NP_776099.1 | 5 | 1787 | ||
| GenBank mRNA | AK132317.1 | - | 4 | 2026 | ||
| AK139326.1 | - | 4 | 2050 | |||
| AK141787.1 | - | 2080 | ||||
| BC027344.1 | - | 5 | 1787 | 3709361 | ||

Zfp940
Genomic Map