siRNA Details
| Gene Symbol: Zfp382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081007.1 | NP_001074476.1 | 5 | 686 | ||
| GenBank mRNA | AK134263.1 | - | 5 | 663 | ||
| BC151163.1 | - | 5 | 621 | 9088052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp382| Gene Symbol: Zfp382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081007.1 | NP_001074476.1 | 5 | 686 | ||
| GenBank mRNA | AK134263.1 | - | 5 | 663 | ||
| BC151163.1 | - | 5 | 621 | 9088052 | ||