siRNA Details
Gene Symbol: Zfp382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081007.1 | NP_001074476.1 | 5 | 486 | ||
GenBank mRNA | AK134263.1 | - | 5 | 463 | ||
BC151163.1 | - | 5 | 421 | 9088052 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081007.1 | NP_001074476.1 | 5 | 486 | ||
GenBank mRNA | AK134263.1 | - | 5 | 463 | ||
BC151163.1 | - | 5 | 421 | 9088052 |