siRNA Details
| Gene Symbol: Alkbh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198027.2 | NP_932144.2 | 6 | 464 | ||
| GenBank mRNA | AK082058.1 | - | 5 | 632 | ||
| AK209174.1 | - | 2 | 112 | |||
| BC029805.1 | - | 6 | 416 | 4952089 | ||
| BC080310.1 | - | 4 | 587 | |||

Alkbh6
Genomic Map