siRNA Details
| Gene Symbol: Scgb2b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100464.2 | NP_001093934.1 | 2 | 293 | ||
| NM_001276475.1 | NP_001263404.1 | 2 | 293 | |||
| GenBank mRNA | AY293278.1 | - | 234 | |||
| AY293279.1 | - | 1 | 148 | |||
| AY293280.1 | - | 1 | 148 | |||
| BC024677.1 | - | 2 | 217 | 4193111 | ||
| BC057695.1 | - | 2 | 293 | |||
| BC127149.1 | - | 234 | 40053058 | |||
| GU269237.1 | - | 2 | 217 | |||

Scgb2b27
Genomic Map