siRNA Details
| Gene Symbol: Kctd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146188.1 | NP_666300.1 | 5 | 832 | ||
| GenBank mRNA | AK034731.1 | - | 6 | 904 | ||
| BC031749.1 | - | 5 | 832 | 4507203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd15| Gene Symbol: Kctd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146188.1 | NP_666300.1 | 5 | 832 | ||
| GenBank mRNA | AK034731.1 | - | 6 | 904 | ||
| BC031749.1 | - | 5 | 832 | 4507203 | ||