siRNA Details
| Gene Symbol: Siglecf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271019.1 | NP_001257948.1 | 3 | 379 | ||
| NM_145581.2 | NP_663556.1 | 3 | 379 | |||
| GenBank mRNA | AF293371.1 | - | 3 | 315 | ||
| AK087658.1 | - | 2 | 442 | |||
| AK146203.1 | - | 2 | 416 | |||
| AK155469.1 | - | 3 | 327 | |||
| AY210401.1 | - | 2 | 275 | |||
| BC145038.1 | - | 2 | 310 | 9053586 | ||
| BC145987.1 | - | 2 | 310 | 40131290 | ||

Siglecf
Genomic Map