siRNA Details
| Gene Symbol: Mrgprb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205795.1 | NP_991364.1 | 1 | 41 | ||
| GenBank mRNA | BC107202.1 | - | 1 | 34 | ||
| BC107203.1 | - | 1 | 34 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrgprb4| Gene Symbol: Mrgprb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205795.1 | NP_991364.1 | 1 | 41 | ||
| GenBank mRNA | BC107202.1 | - | 1 | 34 | ||
| BC107203.1 | - | 1 | 34 | |||
Set Membership: |
|