siRNA Details
| Gene Symbol: PNLIPRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303135.1 | NP_001290064.1 | 4 | 262 | ||
| NM_006229.3 | NP_006220.1 | 4 | 272 | |||
| XM_011539868.1 | XP_011538170.1 | 4 | 405 | |||
| XM_011539869.1 | XP_011538171.1 | 4 | 259 | |||
| GenBank mRNA | AK130236.1 | - | 4 | 260 | ||
| AK307063.1 | - | 4 | 259 | |||
| BC005233.1 | - | 4 | 272 | |||
| BC025784.2 | - | 3 | 235 | 5226231 | ||
| BP323340.1 | - | 4 | 262 | |||
| CR749299.1 | - | 4 | 271 | |||
| CR749524.1 | - | 4 | 271 | |||
| CR749596.1 | - | 4 | 273 | |||
| M93283.1 | - | 4 | 254 | |||

PNLIPRP1
Genomic Map