siRNA Details
| Gene Symbol: Vmn2r66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033878.3 | NP_001029050.3 | 3 | 756 | ||
| GenBank mRNA | AK156873.1 | - | 1 | 677 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r66| Gene Symbol: Vmn2r66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033878.3 | NP_001029050.3 | 3 | 756 | ||
| GenBank mRNA | AK156873.1 | - | 1 | 677 | ||
Set Membership: |
|