siRNA Details
| Gene Symbol: PNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002686.4 | NP_002677.1 | 3 | 916 | ||
| NR_073461.1 | 3 | 781 | ||||
| XM_011524909.2 | XP_011523211.1 | 3 | 1024 | |||
| GenBank mRNA | BC037246.1 | - | 3 | 1115 | 5175571 | |
| J03727.1 | - | 3 | 899 | |||

PNMT
Genomic Map