siRNA Details
| Gene Symbol: P2ry6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183168.2 | NP_898991.1 | 3 | 956 | ||
| GenBank mRNA | AK142680.1 | - | 3 | 981 | ||
| AX775901.1 | - | 3 | 916 | |||
| BC027331.1 | - | 3 | 952 | 3982042 | ||
| BC064095.1 | - | 3 | 936 | 1054566 | ||
More Information
Set Membership: |
|

P2ry6
Genomic Map