siRNA Details
Gene Symbol: Olfr553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207621.1 | NP_997504.1 | 1 | 426 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
