siRNA Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173767.3 | NP_776128.2 | 6 | 890 | ||
| GenBank mRNA | AK014452.1 | - | 5 | 651 | ||
| AK050143.1 | - | 3 | 385 | |||
| AK179765.1 | - | 4 | 273 | |||
| AK187129.1 | - | 2 | 160 | |||
| BC026405.1 | - | 6 | 777 | 4211657 | ||
| BC089586.1 | - | 3 | 394 | |||
| BC139196.1 | - | 3 | 278 | 8862218 | ||
| BC139197.1 | - | 3 | 278 | 8862219 | ||
| DQ205645.1 | - | 5 | 603 | |||

Insc
Genomic Map