siRNA Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173767.3 | NP_776128.2 | 11 | 1599 | ||
| GenBank mRNA | AK050143.1 | - | 8 | 1094 | ||
| AK178410.1 | - | 3 | 273 | |||
| AK196619.1 | - | 3 | 273 | |||
| BC026405.1 | - | 11 | 1486 | 4211657 | ||
| BC139196.1 | - | 8 | 987 | 8862218 | ||
| BC139197.1 | - | 8 | 987 | 8862219 | ||
| DQ205645.1 | - | 10 | 1312 | |||

Insc
Genomic Map