siRNA Details
| Gene Symbol: Plekha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040271.1 | - | 23 | 3374 | ||
| AK146866.1 | - | 3565 | ||||
| BC060678.1 | - | 22 | 3295 | |||
| EU380771.1 | - | 3591 | ||||
| EU380772.1 | - | 3649 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekha7| Gene Symbol: Plekha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040271.1 | - | 23 | 3374 | ||
| AK146866.1 | - | 3565 | ||||
| BC060678.1 | - | 22 | 3295 | |||
| EU380771.1 | - | 3591 | ||||
| EU380772.1 | - | 3649 | ||||