siRNA Details
| Gene Symbol: SEPT5-GP1BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037611.1 | 6 | 2082 | |||
| NR_037612.1 | 6 | 586 | ||||
| GenBank mRNA | AB209168.1 | - | 6 | 2082 | ||
| U59632.1 | - | 6 | 586 | |||
| Gene Symbol: SEPT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009939.2 | NP_001009939.1 | 6 | 867 | ||
| NM_002688.5 | NP_002679.2 | 7 | 662 | |||
| GenBank mRNA | AK056273.1 | - | 6 | 844 | ||
| AK295940.1 | - | 7 | 531 | |||
| BC025261.1 | - | 7 | 626 | 4622405 | ||
| CR456545.1 | - | 7 | 542 | |||
| KJ534756.1 | - | 7 | 537 | |||
| U74628.1 | - | 7 | 624 | |||
| U85431.1 | - | 7 | 603 | |||
| Y11593.1 | - | 7 | 582 | |||

SEPT5-GP1BB
Genomic Map