siRNA Details
| Gene Symbol: Cog7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033318.3 | NP_001028490.2 | 3 | 514 | ||
| GenBank mRNA | AK030709.1 | - | 3 | 481 | ||
| AK145163.1 | - | 3 | 512 | |||
| AK154693.1 | - | 3 | 516 | |||
| AK163143.1 | - | 534 | ||||
| AK172200.1 | - | 3 | 516 | |||
| AK202651.1 | - | 2 | 105 | |||

Cog7
Genomic Map