siRNA Details
| Gene Symbol: Tnrc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144925.3 | NP_659174.3 | 6 | 2245 | ||
| GenBank mRNA | AK083848.1 | - | 5 | 1850 | ||
| AK147327.1 | - | 6 | 1950 | |||
| BC026612.1 | - | 1 | 1669 | |||
| BC026849.1 | - | 6 | 1918 | |||
| CD802917.1 | - | 1 | 60 | |||

Tnrc6a
Genomic Map