siRNA Details
| Gene Symbol: Tnrc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144925.3 | NP_659174.3 | 12 | 4219 | ||
| GenBank mRNA | AK138426.1 | - | 1 | 122 | ||
| AK147327.1 | - | 12 | 3924 | |||
| BC005741.1 | - | 1 | 54 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnrc6a| Gene Symbol: Tnrc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144925.3 | NP_659174.3 | 12 | 4219 | ||
| GenBank mRNA | AK138426.1 | - | 1 | 122 | ||
| AK147327.1 | - | 12 | 3924 | |||
| BC005741.1 | - | 1 | 54 | |||