siRNA Details
| Gene Symbol: D430042O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081022.1 | NP_001074491.1 | 14 | 1774 | ||
| GenBank mRNA | AK052492.1 | - | 1 | 172 | ||
| AK052528.1 | - | 9 | 1281 | |||
| BC157963.1 | - | 13 | 1706 | |||
| BC158019.1 | - | 14 | 1762 | 9007325 | ||

D430042O09Rik
Genomic Map