siRNA Details
| Gene Symbol: Tufm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163713.1 | NP_001157185.1 | 3, 4 | 492 | ||
| NM_172745.3 | NP_766333.1 | 3, 4 | 492 | |||
| GenBank mRNA | AK075857.1 | - | 3, 4 | 416 | ||
| AK084724.1 | - | 3, 4 | 473 | |||
| AK152858.1 | - | 3, 4 | 487 | |||
| AK153135.1 | - | 3, 4 | 487 | |||
| BC060959.1 | - | 3, 4 | 469 | 30379757 | ||
| BC100596.1 | - | 3, 4 | 485 | 6762419 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106295, XM_006230225, XM_006230224.

Tufm
Genomic Map