siRNA Details
| Gene Symbol: Hirip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172746.3 | NP_766334.1 | 3 | 760 | ||
| GenBank mRNA | AK045122.1 | - | 3 | 295 | ||
| AK135526.1 | - | 3 | 303 | |||
| AK161657.1 | - | 3 | 295 | |||
| BC055687.1 | - | 3 | 760 | 6414655 | ||

Hirip3
Genomic Map