siRNA Details
| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172747.2 | NP_766335.1 | 6 | 953 | ||
| GenBank mRNA | AF534881.3 | - | 6 | 939 | ||
| AK075722.1 | - | 6 | 954 | |||
| BC079607.1 | - | 6 | 946 | 30359035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd13| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172747.2 | NP_766335.1 | 6 | 953 | ||
| GenBank mRNA | AF534881.3 | - | 6 | 939 | ||
| AK075722.1 | - | 6 | 954 | |||
| BC079607.1 | - | 6 | 946 | 30359035 | ||