siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172281.2 | NP_758485.2 | 7, 8 | 1112 | ||
| GenBank mRNA | AK089805.1 | - | 7, 8 | 1044 | ||
| AK129184.1 | - | 7, 8 | 1030 | |||
| AK154982.1 | - | 7, 8 | 1043 | |||
| AK171462.1 | - | 7, 8 | 1041 | |||
| AK178704.1 | - | 1, 2 | 170 | |||
| BC038348.1 | - | 7, 8 | 1026 | 4187484 | ||

Rnf40
Genomic Map