siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172281.2 | NP_758485.2 | 15 | 2328 | ||
| GenBank mRNA | AK042654.1 | - | 6 | 988 | ||
| AK089805.1 | - | 15 | 2260 | |||
| AK129184.1 | - | 15 | 2246 | |||
| AK154982.1 | - | 15 | 2259 | |||
| AK171462.1 | - | 15 | 2257 | |||
| BC038348.1 | - | 15 | 2242 | 4187484 | ||

Rnf40
Genomic Map