siRNA Details
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172749.4 | NP_766337.2 | 2 | 963 | ||
| GenBank mRNA | AK078520.1 | - | 2 | 686 | ||
| BC068300.1 | - | 1 | 566 | 30105973 | ||
| BC079907.1 | - | 2 | 963 | 30647214 | ||
| BC098458.1 | - | 1178 | ||||

Zfp646
Genomic Map