siRNA Details
| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139149.2 | NP_631888.1 | 12, 13 | 1351 | ||
| GenBank mRNA | AF224264.1 | - | 12, 13 | 1276 | ||
| AK084867.1 | - | 5, 6 | 2134 | |||
| AK132141.1 | - | 2693 | ||||
| AK139960.1 | - | 12, 13 | 1350 | |||
| AK145582.1 | - | 12, 13 | 1341 | |||
| AK146198.1 | - | 12, 13 | 1344 | |||
| BC011078.1 | - | 6, 7 | 620 | |||
| BC040827.1 | - | 12, 13 | 1306 | 4503820 | ||
| BC058247.1 | - | 12, 13 | 1326 | 6314291 | ||

Fus
Genomic Map