siRNA Details
| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139149.2 | NP_631888.1 | 10, 11 | 1126 | ||
| GenBank mRNA | AF224264.1 | - | 10, 11 | 1051 | ||
| AK084867.1 | - | 3, 4 | 1908 | |||
| AK132141.1 | - | 1119 | ||||
| AK139960.1 | - | 10, 11 | 1125 | |||
| AK145582.1 | - | 10, 11 | 1116 | |||
| AK146198.1 | - | 10, 11 | 1119 | |||
| BC040827.1 | - | 10, 11 | 1081 | 4503820 | ||
| BC058247.1 | - | 10, 11 | 1101 | 6314291 | ||

Fus
Genomic Map