siRNA Details
| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146206.4 | NP_666318.2 | 16, 17 | 1505 | ||
| GenBank mRNA | AK052930.1 | - | 16, 17 | 1506 | ||
| BC043472.1 | - | 12, 13 | 1083 | |||
| BC141195.1 | - | 16, 17 | 1589 | 9055807 | ||

Tpcn2
Genomic Map