siRNA Details
| Gene Symbol: Tpte | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199257.2 | NP_954866.2 | 3, 4 | 483 | ||
| GenBank mRNA | AJ311311.1 | - | 3, 4 | 486 | ||
| AJ311313.1 | - | 3, 4 | 486 | |||
| BC107329.2 | - | 3, 4 | 278 | 40054415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpte| Gene Symbol: Tpte | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199257.2 | NP_954866.2 | 3, 4 | 483 | ||
| GenBank mRNA | AJ311311.1 | - | 3, 4 | 486 | ||
| AJ311313.1 | - | 3, 4 | 486 | |||
| BC107329.2 | - | 3, 4 | 278 | 40054415 | ||