siRNA Details
| Gene Symbol: Sorbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205219.1 | NP_001192148.1 | 23 | 3852 | ||
| NM_172752.4 | NP_766340.4 | 22 | 3798 | |||
| GenBank mRNA | AK122369.1 | - | 18 | 3263 | ||
| AK140498.1 | - | 18 | 2037 | |||
| BC031203.1 | - | 6 | 571 | |||
| BC151038.1 | - | 23 | 3722 | 9087951 | ||
| BC157944.1 | - | 19 | 1943 | 9007279 | ||
| BC172140.1 | - | 22 | 3668 | 9054810 | ||

Sorbs2
Genomic Map