siRNA Details
| Gene Symbol: Sorbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205219.1 | NP_001192148.1 | 21 | 3741 | ||
| NM_172752.4 | NP_766340.4 | 20 | 3687 | |||
| GenBank mRNA | AK050951.1 | - | 16 | 1962 | ||
| AK083429.1 | - | 18 | 2021 | |||
| AK122369.1 | - | 16 | 3152 | |||
| AK140498.1 | - | 16 | 1926 | |||
| BC151038.1 | - | 21 | 3611 | 9087951 | ||
| BC157944.1 | - | 17 | 1832 | 9007279 | ||
| BC172140.1 | - | 20 | 3557 | 9054810 | ||

Sorbs2
Genomic Map