siRNA Details
| Gene Symbol: Sorbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205219.1 | NP_001192148.1 | 15 | 1446 | ||
| NM_172752.4 | NP_766340.4 | 14 | 1389 | |||
| GenBank mRNA | AK050951.1 | - | 11 | 1245 | ||
| AK083429.1 | - | 13 | 1307 | |||
| AK122369.1 | - | 10 | 857 | |||
| AK139388.1 | - | 13 | 1335 | |||
| AK140498.1 | - | 11 | 1209 | |||
| BC039163.1 | - | 15 | 1377 | |||
| BC151038.1 | - | 15 | 1316 | 9087951 | ||
| BC157944.1 | - | 12 | 1118 | 9007279 | ||
| BC172140.1 | - | 14 | 1259 | 9054810 | ||

Sorbs2
Genomic Map