siRNA Details
| Gene Symbol: Ddx60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293783.1 | NP_001280712.1 | 36 | 5237 | ||
| GenBank mRNA | AK136403.1 | - | 3 | 2317 | ||
| BC013672.1 | - | 9 | 1225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx60| Gene Symbol: Ddx60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293783.1 | NP_001280712.1 | 36 | 5237 | ||
| GenBank mRNA | AK136403.1 | - | 3 | 2317 | ||
| BC013672.1 | - | 9 | 1225 | |||