siRNA Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145597.4 | NP_663572.1 | 2, 3 | 145 | ||
| GenBank mRNA | AK049979.1 | - | 2, 3 | 144 | ||
| AK082758.1 | - | 2, 3 | 141 | |||
| AK147962.1 | - | 2, 3 | 138 | |||
| BC021367.1 | - | 2, 3 | 145 | 5030434 | ||

Tmem161a
Genomic Map