siRNA Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024617.3 | NP_001019788.1 | 7 | 837 | ||
| NM_001297591.1 | NP_001284520.1 | 6 | 763 | |||
| NM_001297593.1 | NP_001284522.1 | 5 | 644 | |||
| NM_001297596.1 | NP_001284525.1 | 6 | 763 | |||
| GenBank mRNA | AK141502.1 | - | 8 | 760 | ||
| BC064813.1 | - | 5 | 539 | 6826283 | ||
| DQ296611.1 | - | 6 | 763 | |||
| DQ296612.1 | - | 5 | 644 | |||
| DQ296613.1 | - | 6 | 763 | |||

Inpp4b
Genomic Map