siRNA Details
| Gene Symbol: Heatr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172757.3 | NP_766345.3 | 13 | 1807 | ||
| GenBank mRNA | AK047782.1 | - | 13 | 1838 | ||
| AK052635.1 | - | 13 | 1834 | |||
| AK134510.1 | - | 13 | 1812 | |||
| AK136424.1 | - | 13 | 1817 | |||
| AK140522.1 | - | 13 | 1825 | |||
| AK163715.1 | - | 13 | 2570 | |||
| AK165541.1 | - | 13 | 1701 | |||
| BC030373.1 | - | 9 | 1152 | |||
| BC048074.1 | - | 2 | 525 | |||
| BC065060.1 | - | 13 | 1807 | 6402217 | ||

Heatr3
Genomic Map